Learn More
Sampling Systems™ Metal Detectable Volumetric Spoons
Product Code. 17574716 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
2.5 mL
Unit Size:
Pack of 4
Product Code. Capacity (Metric) unitSize
17574716 2.5 mL Pack of 4
1 options
Product Code. 17574716 Supplier Sampling Systems™ Supplier No. M1030B25
Only null left
Add to basket
Add to basket
Metal Detectable Volumetric Spoons are ideal for general purpose measuring. They are ideal for both powders and liquids. The robust metal detectable PP means they will give a long service life.
  • Metal Detectable Volumetric Spoons are made from a special chemically resistant grade of PP.
  • The whole range is manufactured in cleanroom conditions and packed in a class 7 (10,000) cleanroom.
  • This ensures the cleanliness of the product.
  • The robust design means they can be used for years and years or used as a single use product straight from the bag.
  • Simply unwrap and use.
  • They are ideal for use with powders, liquids and granules.
  • The blue color means they can easily seen if left in the powder in line with HACCP guidelines.
  • Made from high quality PP.
  • Conforms to the latest FDA and EU food requirements.
  • 2.5 mL volume is a standard size.
Specifications
Capacity (Metric) 2.5 mL
Color Blue
Material Polypropylene