Learn More
Sampling Systems™ Metal Detectable Long Handled Spoons
Product Code. 17594716 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
20 mL
Unit Size:
Pack of 4
Product Code. Capacity (Metric) unitSize
17594716 20 mL Pack of 4
1 options
Product Code. 17594716
Brand: Sampling Systems™ M1036B20
Only null left
Add to basket
Add to basket
Metal Detectable Long Handled Spoons with long handles make taking powder and liquid samples easy. They can also be used for general production work. The special metal detectable PP means the material will be picked up by a ray and general detection equipment.
  • Powder sampling is easy with our metal detectable spoons.
  • They have a very long handle which means they are easy to see if left in the powder.
  • They are made from a special robust, chemically resistant grade of PP.
  • The whole range is manufactured in cleanroom conditions and packed in a class 7 (10,000) cleanroom.
  • This ensures the cleanliness of the product.
  • The robust design means they can be used for years and years or used as a single use product straight from the bag.
  • Simply unwrap and use.
  • They are ideal for use with liquids, creams and pastes.
  • The blue color means they can easily seen if left in the material in line with HACCP guidelines.
  • Made from high quality PP.
  • Conforms to the latest FDA and EU food requirements.
  • 20 mL volume is a standard size.
Specifications
Capacity (Metric) 20 mL
Color Blue
Material Polypropylene