Learn More
Sampling Systems™ Metal Detectable Long Handled Scoops
Product Code. 17504726 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
100 mL
Unit Size:
Pack of 4
Product Code. Capacity (Metric) unitSize
17504726 100 mL Pack of 4
1 options
Product Code. 17504726 Supplier Sampling Systems™ Supplier No. M1040B100
Only null left
Add to basket
Add to basket
Metal Detectable Long Handled Scoops make production work easy. The long handle is ideal for when you want to reach into difficult to reach places. The special metal detectable PP means the material will be picked up by X-ray and general detection equipment.
  • Our Long Handled Metal Detectable Scoops are perfect for both sampling and dispensing when working with powders and granules.
  • The long handle and the bright blue color make the scoops easy to spot if they get left behind.
  • To ensure the cleanliness of the scoops they are manufactured and packed in a Cleanroom.
  • The robust nature of these scoops means that they can give many years of service or, for more demanding applications, they can be used as a single use disposable product straight from the bag.
  • The scoops are made from polypropylene that has been mixed with a special food grade filler.
  • It is the filler that makes the scoops metal detectable and X-Ray visible.
  • The filler is uniformly distributed throughout the scoop so even small fragments of scoop can still be detected.
  • Simply unwrap and use.
  • These scoops are ideal for use with powders, granules and pastes.
  • Our Metal Detectable Long Handle Scoops are designed to be as easy to clean as possible.
  • The main body of the scoop has rounded corners.
  • Made from food grade PP.
  • Conforms to the latest FDA and EU requirements.
Specifications
Capacity (Metric) 100 mL
Color Blue
Material Polypropylene