missing translation for 'onlineSavingsMsg'
Learn More

Sampling Systems™ Metal Detectable Jugs

Product Code. 17514726 Shop All Sampling Systems Products
Change view
Click to view available options
Capacity (Metric):
500 mL
1000 mL
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17514726 1000 mL Pack of 4
17524726 500 mL Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17514726 Supplier Sampling Systems™ Supplier No. M1044B1000
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Metal Detectable Jugs are available in 500 mL and 1000 mL sizes. The robust, chemically resistant PP means the jugs are sturdy and easy to use. The graduations are on the outside so it is simple measure out the liquid.

  • The Sampling Systems range of detectable jugs are ideal for use in food, pharma and chemical industries.
  • They are made from a special chemically resistant grade of PP.
  • The whole range is manufactured in cleanroom conditions and packed in a class 7 (10,000) cleanroom.
  • This ensures the cleanliness of the product.
  • The robust design means they can be used for years and years or used as a single use product straight from the bag.
  • Simply unwrap and use.
  • They are ideal for use with powders, granules and pastes.
  • The blue color means they can easily seen if left in the material.
  • Made from high quality PP.
  • Conforms to the latest FDA and EU food requirements.
  • 500 mL and 1000 mL volume is a standard size.

Specifications

Capacity (Metric) 1000 mL
Color Blue
Material Polypropylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.