Learn More
DWK Life Sciences Metal Cap
Product Code. 11776115 Shop All DWK Life Sciences Products
Click to view available options
Material:
Anodized Aluminum
Stainless Steel
Unit Size:
Pack of 10
Product Code. 11776115
Brand: DWK Life Sciences 290132407
Only null left
Add to basket
Add to basket
The metal cap is used to attach to a culture flask with a straight neck and thus to close it.
Microbiology is an important field of biology and medicine and deals with microorganisms (bacteria, fungi, protozoa or viruses) and their habitats and conditions. For this purpose, it is essential to produce pure culture media on which the corresponding microorganisms can grow. In order to avoid the introduction of foreign substances, the bottles and flasks must be covered but not sealed gas-tight. Air exchange is essential for culture growth.
  • The metal caps allow an exchange of air in the media bottle, creating a positive environment for the growth of microorganisms
  • These caps are autoclavable, but not dishwasher-safe
TRUSTED_SUSTAINABILITY
Specifications
Color Blue
For Use With (Application) Closuring of culture tubes or bottles; Closuring of culture media bottles; Storage and transport of Liquid cultures; Working with cell culture media under sterile conditions in biosafety cabinets and clean hoods. Not dishwasher-safe.
Material Anodized Aluminum
Neck ID (Metric) 38 mm
Quantity 10/Pk.
Diameter (Metric) 38 mm
Autoclavable Autoclavable