missing translation for 'onlineSavingsMsg'
Learn More

Mercury(II) Cyanide, TRC

Product Code. 30180106
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30180106 5 g 5g
30180349 1 g 1g
30179940 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30180106 Supplier Toronto Research Chemicals Supplier No. TRCM2255555G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-04-1
Molecular Formula C2HgN2
Molecular Weight (g/mol) 252.62
Synonym Mercury Cyanide, Cianurina, Dicyanomercury, Mercuric Cyanide, Mercuric Cyanide, Mercury Dicyanide;
IUPAC Name mercury(2+);dicyanide
SMILES [Hg+2].[C-]#N.[C-]#N

Specifications

Quantity 5 g
InChI Formula InChI=1S/2CN.Hg/c2*1-2;/q2*-1;+2
Formula Weight 253.9768
Chemical Name or Material Mercury(II) Cyanide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.