CAS RN 592-04-1
IUPAC Name:
mercurydicarbonitrile
Synonyms:
mercuric cyanide mercury(II) cyanide mercurius cyanatus hydrargyrum cyanatum mercury cyanide quecksilber(II)-cyanid cyanure de mercure Hg(CN)2
Molecular Weight (g/mol):
252.63
Molecular Formula:
C2HgN2
InChi Key:
FQGYCXFLEQVDJQ-UHFFFAOYSA-N
SMILES:
N#C[Hg]C#N
1
–
1
of
1
results
1
Mercury(II) Cyanide, TRC
CAS: 592-04-1 Molecular Formula: C2HgN2 Molecular Weight (g/mol): 252.62 Synonym: Mercury Cyanide,Cianurina,Dicyanomercury,Mercuric Cyanide,Mercuric Cyanide,Mercury Dicyanide; IUPAC Name: mercury(2+);dicyanide SMILES: [Hg+2].[C-]#N.[C-]#N