missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Mer Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
£169.00 - £522.00
Specifications
| Antigen | Mer |
|---|---|
| Clone | 125508 |
| Dilution | Western Blot 1 ug/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18752869
![]() |
R&D Systems
MAB8911-SP |
25 μg |
£169.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18112671
![]() |
R&D Systems
MAB8911 |
500 μg |
£522.00
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mer Monoclonal specifically detects Mer in Human samples. It is validated for Western Blot.Specifications
| Mer | |
| Western Blot 1 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
| MERTK | |
| IgG2b | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 125508 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q12866.2 | |
| 10461 | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Mer Ala21-Ala499 Accession # Q12866.2 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human Mer in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) Axl, rhDtk, or rmMer. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title