missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Mer Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB8911-SP
This item is not returnable.
View return policy
Description
Mer Monoclonal specifically detects Mer in Human samples. It is validated for Western Blot.
Specifications
| Mer | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| c-Eyk, C-mer, c-mer proto-oncogene tyrosine kinase, EC 2.7.10, EC 2.7.10.1, MER, MER receptor tyrosine kinase, MerTK, MGC133349, Receptor tyrosine kinase MerTK, RP38Proto-oncogene c-Mer, STK kinase, tyrosine-protein kinase Mer | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 10461 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot | |
| 125508 | |
| Western Blot 1 ug/mL | |
| Q12866.2 | |
| MERTK | |
| S. frugiperda insect ovarian cell line Sf 21-derived recombinant human Mer Ala21-Ala499 Accession # Q12866.2 | |
| 25 μg | |
| Primary | |
| Detects human Mer in direct ELISAs and Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) Axl, rhDtk, or rmMer. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction