missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mena, Mouse anti-Human, Clone: ENAH/1988, Novus Biologicals™
Mouse Monoclonal Antibody
£203.00 - £377.00
Specifications
| Antigen | Mena |
|---|---|
| Clone | ENAH/1988 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 ug/ml, ELISA, Protein Array |
| Applications | Western Blot, ELISA, Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18222097
|
Novus Biologicals
NBP2-79752-20UG |
20 μg |
£203.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18260098
|
Novus Biologicals
NBP2-79752-100UG |
100 μg |
£377.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mena Monoclonal specifically detects Mena in Human samples. It is validated for Western Blot, ELISA, Protein Array.Specifications
| Mena | |
| 0.2 mg/mL | |
| Western Blot, ELISA, Peptide Array | |
| Unconjugated | |
| Mouse | |
| Cancer, Neuroscience | |
| 10 mM PBS with 0.05% BSA | |
| ENA, enabled homolog (Drosophila), FLJ10773, MENANDPP1, protein enabled homolog | |
| ENAH | |
| IgG2c κ | |
| Protein A or G purified |
| ENAH/1988 | |
| Western Blot 1-2 ug/ml, ELISA, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Q8N8S7, Q8N8S7 | |
| 55740 | |
| Recombinant fragment of human Mena protein (around aa 485-589) (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title