missing translation for 'onlineSavingsMsg'
Learn More

Mena, Mouse anti-Human, Clone: ENAH/1988, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP2-79752-100UG

 View more versions of this product

Product Code. 18260098

  • £377.00 / 100µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Mena Monoclonal specifically detects Mena in Human samples. It is validated for Western Blot, ELISA, Protein Array.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Mena
Monoclonal
0.2 mg/mL
Western Blot 1-2 ug/ml, ELISA, Protein Array
Q8N8S7, Q8N8S7
ENAH
Recombinant fragment of human Mena protein (around aa 485-589) (exact sequence is proprietary)
100 μg
Cancer, Neuroscience
55740
Store at 4°C.
IgG2c κ
Western Blot, ELISA, Peptide Array
ENAH/1988
Unconjugated
10 mM PBS with 0.05% BSA
ENA, enabled homolog (Drosophila), FLJ10773, MENANDPP1, protein enabled homolog
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.