missing translation for 'onlineSavingsMsg'
Learn More

Nabertherm™ Melting Furnace R7 Controller

These compact melting furnaces for the melting of non-ferrous metals and alloys are one of a kind and have a number of technical advantages. Designed as tabletop models, they can be used for many laboratory applications

Brand:  Nabertherm™ KD0130IRNA


Code : NEW

Additional Details : Weight : 125.00000kg

 View more versions of this product

Product Code. 15531012

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.


Description

Description

  • Controller: R7
  • tabletop model
  • Electrical connection: 1-phase
Specifications

Specifications

R7
Crucible A 6
520 x 680 x 660 mm
0.75 L
1300°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers