missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Melanoma Associated Antigen (KBA.62) Antibody (KBA.62), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-45285-0.02MG
This item is not returnable.
View return policy
Description
Ensure accurate, reproducible results in Immunohistochemistry (Paraffin), Immunofluorescence
Melanoma Associated Antigen (KBA.62) Monoclonal specifically detects Melanoma Associated Antigen (KBA.62) in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunofluorescence.
Specifications
| Melanoma Associated Antigen (KBA.62) | |
| Monoclonal | |
| 0.2mg/mL | |
| Immunohistochemistry-Paraffin 0.5 - 1.0 ug/ml, SDS-Page, Immunofluorescence 0.5 - 1.0 ug/ml | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| KBA.62 is a novel anti-melanoma antibody. It reacts positively against melanocytic tumors but not other tumors, thus demonstrating specificity and sensitivity. Moreover, it reacts positively against junctional nevus cells but not intradermal nevi, and against fetal melanocytes but not normal adult melanocytes. KBA.62 antibody is useful in identifying malignant melanomas. Metastatic amelanotic melanoma can often be confused with a variety of poorly differentiated carcinomas, large cell lymphomas, sarcomas, spindle cell carcinomas and various types of mesenchymal neoplasms. A keratin-negative, vimentin-rich neoplasm that immuno-reacts with antibody to S-100 protein and with KBA.62 antibody is, with rare exception, a melanoma. Anti-KBA.62 is a useful additional marker for melanoma, specifically in desmoplastic/spindle cell cases and in the context of micro-metastasis in sentinel lymph node. | |
| Store at 4C. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin), SDS-Page, Immunofluorescence | |
| KBA.62 | |
| Unconjugated | |
| 10mM PBS and 0.05% BSA with 0.05% Sodium Azide | |
| Human KAL cells derived from lymph node metastasis of malignant melanoma | |
| 0.02 mg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction