missing translation for 'onlineSavingsMsg'
Learn More

MEKK3 Antibody (2H11), Novus Biologicals™

Rabbit Monoclonal Antibody

£260.00 - £406.00

Specifications

Antigen MEKK3
Clone 2H11
Dilution ELISA
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30033456
View Documents
Novus Biologicals
NBP3-26151-50ul
50 μL
£260.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30035929
View Documents
Novus Biologicals
NBP3-26151-100ul
100 μL
£406.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MEKK3 Monoclonal antibody specifically detects MEKK3 in Human samples. It is validated for ELISA
Specifications

Specifications

MEKK3
ELISA
Monoclonal
Purified
RUO
Human
EC 2.7.11.25, MAP/ERK kinase kinase 3, MAPK/ERK kinase kinase 3, MAPKKK3MEKK 3, MEK kinase 3, MEKK3, mitogen-activated protein kinase kinase kinase 3
A synthesized peptide derived from Human MEKK3 [UniProt Q99759]
Primary
Store at -20 to -70C. Avoid freeze-thaw cycles.
2H11
ELISA
Unconjugated
Rabbit
Angiogenesis, Cancer, Immunology, Innate Immunity, Protein Kinase
PBS, pH 7.4, 150mM NaCl and 50% glycerol
4215
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.