missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEKK3 Antibody (2H11), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-26151-100ul
This item is not returnable.
View return policy
Description
MEKK3 Monoclonal antibody specifically detects MEKK3 in Human samples. It is validated for ELISA
Specifications
| MEKK3 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl and 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 2H11 | |
| ELISA | |
| EC 2.7.11.25, MAP/ERK kinase kinase 3, MAPK/ERK kinase kinase 3, MAPKKK3MEKK 3, MEK kinase 3, MEKK3, mitogen-activated protein kinase kinase kinase 3 | |
| A synthesized peptide derived from Human MEKK3 [UniProt Q99759] | |
| 100 μL | |
| Angiogenesis, Cancer, Immunology, Innate Immunity, Protein Kinase | |
| 4215 | |
| Store at -20 to -70C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction