missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEIS2 Rabbit anti-Human, Mouse, Clone: 2A6I3, Novus Biologicals™
Rabbit Monoclonal Antibody
£150.00 - £386.00
Specifications
| Antigen | MEIS2 |
|---|---|
| Clone | 2A6I3 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18369334
|
Novus Biologicals
NBP3-16224-20UL |
20 μg |
£150.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18310702
|
Bio-Techne
NBP3-16224-100UL |
100 μg |
£386.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MEIS2 Monoclonal antibody specifically detects MEIS2 in Human, Mouse samples. It is validated for Western BlotSpecifications
| MEIS2 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Vision | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| 4212 | |
| IgG | |
| Affinity purified |
| 2A6I3 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Meis (mouse) homolog 2, Meis homeobox 2, Meis1, myeloid ecotropic viral integration site 1 homolog 2 (mouse), Meis1-related gene 1, MGC2820, myeloid ecotropic viral integration site 1 homolog 2, TALE homeobox protein Meis2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MEIS2 (O14770). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title