missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MEIS2 Rabbit anti-Human, Mouse, Clone: 2A6I3, Novus Biologicals™
Description
MEIS2 Monoclonal antibody specifically detects MEIS2 in Human, Mouse samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | MEIS2 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 2A6I3 |
| Conjugate | Unconjugated |
| Formulation | PBS, 0.05% BSA, 50% glycerol, pH7.3 |
| Gene Alias | Meis (mouse) homolog 2, Meis homeobox 2, Meis1, myeloid ecotropic viral integration site 1 homolog 2 (mouse), Meis1-related gene 1, MGC2820, myeloid ecotropic viral integration site 1 homolog 2, TALE homeobox protein Meis2 |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide corresponding to a sequence within amino acids 1-100 of human MEIS2 (O14770). |
| Purification Method | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?