Learn More
KD Scientific Medium Pressure Nut/Ferrule Auto-fill Kit
Product Code. 13423659
Click to view available options
IP Rating:
None
Unit Size:
Each
Product Code. 13423659
Brand: KD Scientific 788347
Only null left
Add to basket
Add to basket
Medium Pressure Nut/Ferrule Auto Fill Kit contains Nut PEEK 1/16, 1/4-28 (3 pcs), Ferrule, flangeless 1/16 (3 pcs) and Teflon tubing (5 ft/1.5 m).
  • Spare part / accessory for KD Scientific Syringe pumps
  • Compatible with KD Legacy and Legato™ products
Specifications
IP Rating None
Type Auto-fill Kit