missing translation for 'onlineSavingsMsg'
Learn More
Learn More
mediator of cell motility 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-54373
This item is not returnable.
View return policy
Description
mediator of cell motility 1 Polyclonal specifically detects mediator of cell motility 1 in Human samples. It is validated for Western Blot.
Specifications
| mediator of cell motility 1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| C21orf19-like protein, C2orf4DKFZp434I0135, CGI-27, FLJ25031, HCV NS5A-transactivated protein 7, Hepatitis C virus NS5A-transactivated protein 7, mediator of cell motility 1, Mediator of ErbB2-driven cell motility 1, memo-1, MEMOchromosome 2 open reading frame 4, NS5ATP7, protein MEMO1 | |
| Rabbit | |
| 34 kDa | |
| 100 μL | |
| Signal Transduction | |
| 51072 | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Zebrafish | |
| IgG |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9Y316 | |
| MEMO1 | |
| Synthetic peptides corresponding to MEMO1(mediator of cell motility 1) The peptide sequence was selected from the middle region of MEMO1. Peptide sequence AMESHKDEFTIIPVLVGALSESKEQEFGKLFSKYLADPSNLFVVSSDFCH. | |
| Affinity purified | |
| RUO | |
| Primary | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction