missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MDMX Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£292.00 - £435.00
Specifications
| Antigen | MDMX |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18429831
|
Novus Biologicals
NBP1-87733-25ul |
25 μL |
£292.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18263696
|
Novus Biologicals
NBP1-87733 |
0.1 mL |
£435.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MDMX Polyclonal specifically detects MDMX in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MDMX | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| DKFZp781B1423, double minute 4 homolog, Double minute 4 protein, double minute 4, human homolog of; p53-binding protein, HDMX, Mdm2-like p53-binding protein, Mdm4 p53 binding protein homolog (mouse), Mdm4, transformed 3T3 cell double minute 4, p53 binding protein, Mdm4, transformed 3T3 cell double minute 4, p53 binding protein (mouse), MDM4-related protein 1, MDMXMGC132766, mouse double minute 4, human homolog of; p53-binding protein, MRP1, p53-binding protein Mdm4, protein Mdm4, Protein Mdmx | |
| MDM4 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Apoptosis, DNA Repair, Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 4194 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SFSVKDPSPLYDMLRKNLVTLATATTDAAQTLALAQDHSMDIPSQDQLKQSAEESSTSRKRTTEDDIPTLP | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title