missing translation for 'onlineSavingsMsg'
Learn More

MDA5 Antibody (1B6), Novus Biologicals™

Mouse Monoclonal Antibody

£163.00 - £439.00

Specifications

Antigen MDA5
Clone 1B6
Dilution Western Blot 1:1000, ELISA
Applications Western Blot, ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18141930
View Documents
Novus Biologicals
NBP1-97526-0.025MG
0.025 mg
£163.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18056296
View Documents
Novus Biologicals
NBP1-97526
0.1 mg
£439.00
0.10mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MDA5 Monoclonal specifically detects MDA5 in Human, Mouse samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

MDA5
Western Blot 1:1000, ELISA
Monoclonal
Purified
RUO
Human, Mouse
64135
Recombinant human MDA5 (melanoma differentiation-associated gene 5) (aa 78-555).
Primary
Store at -20C. Avoid freeze-thaw cycles.
1B6
Western Blot, ELISA
Unconjugated
Mouse
Cell Cycle and Replication
CADM-140 autoantigen, Clinically amyopathic dermatomyositis autoantigen 140 kDa, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, EC 3.6.4.13, Helicard, Helicase with 2 CARD domains, Hlcd, IDDM19, interferon induced with helicase C domain 1, interferon-induced helicase C domain-containing protein 1, Interferon-induced with helicase C domain protein 1, MDA-5MGC133047, MDA5RNA helicase-DEAD box protein 116, melanoma differentiation associated protein-5, Melanoma differentiation-associated protein 5, Murabutide down-regulated protein, RH116
IFIH1
IgG1
Protein G purified
50 kDa
Videos
SDS
Documents

Documents

Certificates

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.