missing translation for 'onlineSavingsMsg'
Learn More

MDA5 Antibody (1B6), Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP1-97526-0.025MG

 View more versions of this product

Product Code. 18141930

  • £163.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MDA5 Monoclonal specifically detects MDA5 in Human, Mouse samples. It is validated for Western Blot, ELISA.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MDA5
Monoclonal
Unconjugated
PBS and 50% Glycerol with 0.09% Sodium Azide
IFIH1
Recombinant human MDA5 (melanoma differentiation-associated gene 5) (aa 78-555).
Protein G purified
RUO
Primary
Human, Mouse
Purified
Western Blot, ELISA
1B6
Western Blot 1:1000, ELISA
CADM-140 autoantigen, Clinically amyopathic dermatomyositis autoantigen 140 kDa, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide, EC 3.6.4.13, Helicard, Helicase with 2 CARD domains, Hlcd, IDDM19, interferon induced with helicase C domain 1, interferon-induced helicase C domain-containing protein 1, Interferon-induced with helicase C domain protein 1, MDA-5MGC133047, MDA5RNA helicase-DEAD box protein 116, melanoma differentiation associated protein-5, Melanoma differentiation-associated protein 5, Murabutide down-regulated protein, RH116
Mouse
50 kDa
0.025 mg
Cell Cycle and Replication
64135
Store at -20C. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only