missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MD-1/LY86 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
£150.00
Specifications
| Antigen | MD-1/LY86 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
MD-1/LY86 Polyclonal specifically detects MD-1/LY86 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MD-1/LY86 | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| Polyclonal | |
| Rabbit | |
| Adaptive Immunity, Apoptosis, Immunology | |
| PBS with 0.02% Sodium Azide | |
| Ly-86, lymphocyte antigen 86, MD1, MMD-1, Protein MD-1, RP105-associated | |
| LY86 | |
| IgG | |
| Affinity Purified |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| NP_004262 | |
| 9450 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the center of human MD-1. The immunogen is located within the last 50 amino acids of MD-1. Amino Acid Squence: CGRRKGEQIYYAGPV | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title