missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MD-1/LY86 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76709-0.025mg
This item is not returnable.
View return policy
Description
MD-1/LY86 Polyclonal specifically detects MD-1/LY86 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MD-1/LY86 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Ly-86, lymphocyte antigen 86, MD1, MMD-1, Protein MD-1, RP105-associated | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 2.5 μg/mL, Immunohistochemistry-Paraffin 2.5 μg/mL | |
| NP_004262 | |
| LY86 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the center of human MD-1. The immunogen is located within the last 50 amino acids of MD-1. Amino Acid Squence: CGRRKGEQIYYAGPV | |
| 0.025 mg | |
| Adaptive Immunity, Apoptosis, Immunology | |
| 9450 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction