missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM7 Mouse, Clone: MCM7/1466, enQuireBio™
Mouse Monoclonal Antibody
£181.83 - £395.00
Specifications
Antigen | MCM7 |
---|---|
Clone | MCM7/1466 |
Concentration | 200μg/mL |
Applications | Western Blot |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15960448
|
EnquireBio
4176-MSM1-P1 |
100 μg |
£395.00
100µg |
Estimated Shipment: 18-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15950448
|
EnquireBio
4176-MSM1-P0 |
20 μg |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Specifications
MCM7 | |
200μg/mL | |
Monoclonal | |
Mouse | |
10mM PBS with 0.05% BSA and 0.05% Azide | |
4176 | |
Recombinant human MCM7 protein fragment (aa195-319) | |
Primary | |
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
88kDa |
MCM7/1466 | |
Western Blot | |
Unconjugated | |
RUO | |
NP_005907 | |
MCM7 | |
IgG2b | |
Protein A/G Purified | |
Nuclear | |
Raji or Ramos cells. Tonsil |