missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MCM6 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Brand: Novus Biologicals NBP1-82642
This item is not returnable.
View return policy
Description
MCM6 Polyclonal specifically detects MCM6 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Specifications
| MCM6 | |
| Polyclonal | |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200-1:500 | |
| DNA replication licensing factor MCM6, EC 3.6.4.12, MCG40308, MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe), MCM6 minichromosome maintenance deficient 6 (MIS5 homolog, S. pombe) (S.cerevisiae), minichromosome maintenance complex component 6, minichromosome maintenance deficient (mis5, S. pombe) 6, minichromosome maintenance deficient 6 homolog, minichromosome maintenance deficient 6 homolog (S. cerevisiae), Mis5, MIS5 homolog, p105MCM | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of human MCM6 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| MCM6 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:VILRAEAVESAQAGDKCDFTGTLIVVPDVSKLSTPGARAETNSRVSGVDGYETEGIRGLRALGVRDLSYRLVFLACCVAPTNPRFGGKELRDEEQTAESIKNQMTVKEWEKVFEMSQDKNLYHNLCTSLFPTIHGNDEVKRGV | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair | |
| 4175 | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction