Learn More
Thermo Scientific™ MCF7 (+hIGF-1) Control Lysate (1 mg/mL)
Thermo Scientific™ MCF7 (+hIGF-1) Control Lysate is a stimulated cell lysate that has been validated for the expression levels and phosphorylation of key targets in the AKT signaling pathway.
Brand: Thermo Scientific™ A37959
Additional Details : Weight : 0.00250kg
Description
Thermo Scientific™ MCF7 (+hIGF-1) Control Lysate is a stimulated cell lysate that has been validated for the expression levels and phosphorylation of key targets in the AKT signaling pathway.
- Validated for the detection and phosphorylation of AKT, PTEN, IRS1, IGF1-R, GSK3a, GSK3b, PRAS40, mTOR, p70S6K, and TSC2 proteins
- Optimized - stimulation and lysis conditions optimized for detection of AKT signaling targets
- Convenient - lysate is ready-to-use for immunoprecipitation, immunoprecipitation to mass spectrometry, and western blotting applications
- Concentration: 1 mg/mL
- Stable for two years at -80°C
Specifications
Mass Spectrometry | |
Dry Ice | |
Frozen Liquid | |
0.5 mL | |
Protein |
Mass Spectrometer | |
Sample Enrichment | |
Thermo Scientific™ | |
Control Lysate | |
Cell Lysate |