missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCAK Recombinant Rabbit Monoclonal Antibody (23GB2730), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
£162.00 - £387.00
Specifications
| Antigen | MCAK |
|---|---|
| Clone | 23GB2730 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089148
![]() |
Invitrogen™
MA552512 |
100 μL |
£387.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089417
|
Invitrogen™
MA552513 |
20 μL |
£162.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MCAK Recombinant Monoclonal antibody specifically detects MCAK in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| MCAK | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q99661 | |
| 11004 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2730 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 4930402F02Rik; CT139; ESTM5; hypothetical protein; KIF2C; kinesin family member 2C; kinesin-like 6; kinesin-like protein 6; Kinesin-like protein KIF2C; kinesin-related protein 2; KNSL6; KRP2; MCAK; Mitotic centromere-associated kinesin; QtsA-16015; RP11-269F19.1; testis tissue sperm-binding protein Li 68n; X83316 | |
| A synthesized peptide derived from human MCAK (61-86AA). | |
| Primary | |
| -20°C | |
| KIF2C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title