missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MCAK Recombinant Rabbit Monoclonal Antibody (23GB2730), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA552512
This item is not returnable.
View return policy
Description
The transport of molecules between the nucleus and the cytoplasm in eukaryotic cells is mediated by the nuclear pore complex (NPC), which consists of 60-100 proteins. Small molecules (up to 70 kD) can pass through the nuclear pore by nonselective diffusion while larger molecules are transported by an active process. The protein encoded by this gene belongs to the importin alpha family, and is involved in nuclear protein import.
Specifications
| MCAK | |
| Recombinant Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q99661 | |
| KIF2C | |
| A synthesized peptide derived from human MCAK (61-86AA). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| 23GB2730 | |
| Unconjugated | |
| KIF2C | |
| 4930402F02Rik; CT139; ESTM5; hypothetical protein; KIF2C; kinesin family member 2C; kinesin-like 6; kinesin-like protein 6; Kinesin-like protein KIF2C; kinesin-related protein 2; KNSL6; KRP2; MCAK; Mitotic centromere-associated kinesin; QtsA-16015; RP11-269F19.1; testis tissue sperm-binding protein Li 68n; X83316 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 11004 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction