missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
£145.00 - £402.00
Specifications
| Antigen | MBP |
|---|---|
| Dilution | Western Blot 1:10000, Immunohistochemistry 1:5000, Immunocytochemistry/ Immunofluorescence 1:2000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18679108
|
Novus Biologicals
NBP2-50035SS |
0.025 mL |
£145.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18659046
|
Novus Biologicals
NBP2-50035 |
0.1 mL |
£402.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MBP Polyclonal antibody specifically detects MBP in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| MBP | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Chicken | |
| Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers | |
| Supplied as concentrated IgY in PBS | |
| 4155 | |
| IgY | |
| IgY purified |
| Western Blot 1:10000, Immunohistochemistry 1:5000, Immunocytochemistry/ Immunofluorescence 1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Pig, Bovine | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| Purified myelin basic protein isolated from bovine brain. [UniProt# P02687] | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title