missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand: Novus Biologicals NBP2-50035SS
This item is not returnable.
View return policy
Description
MBP Polyclonal antibody specifically detects MBP in Human, Mouse, Rat, Porcine, Bovine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| MBP | |
| Polyclonal | |
| Western Blot 1:10000, Immunohistochemistry 1:5000, Immunocytochemistry/ Immunofluorescence 1:2000 | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| Purified myelin basic protein isolated from bovine brain. [UniProt# P02687] | |
| 0.025 mL | |
| Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers | |
| 4155 | |
| Store at 4°C. | |
| IgY |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Supplied as concentrated IgY in PBS | |
| Chicken | |
| IgY purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction