missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody (MBP14) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
£169.00 - £376.00
Specifications
| Antigen | MBP |
|---|---|
| Clone | MBP14 |
| Dilution | Western Blot 1:500, ELISA 1:2000, Immunohistochemistry 1:100,000, Immunocytochemistry/Immunofluorescence 1:50 |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18203065
|
Novus Biologicals
NBP2-61938 |
0.1 mg | |||||||
|
18625217
|
Novus Biologicals
NBP2-61938-0.025mg |
0.025 mg |
£169.00
0.03mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MBP Monoclonal specifically detects MBP in Human, Bovine, Chicken, Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
| MBP | |
| Western Blot 1:500, ELISA 1:2000, Immunohistochemistry 1:100,000, Immunocytochemistry/Immunofluorescence 1:50 | |
| Monoclonal | |
| Purified | |
| Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers | |
| PBS with 0.02% Sodium Azide | |
| 4155 | |
| Reacts with human MBP, recognizing epitopes in the 36-50 amino acid region (GILDSIGRFFGGDRG) | |
| Primary |
| MBP14 | |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| Unconjugated | |
| Rat | |
| Bovine, Chicken, Equine, Human | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| MBP | |
| IgG2b | |
| Protein G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title