missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MBP Antibody (MBP14) - Azide and BSA Free, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals NBP2-61938-0.025mg
This item is not returnable.
View return policy
Description
MBP Monoclonal specifically detects MBP in Human, Bovine, Chicken, Equine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| MBP | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500, ELISA 1:2000, Immunohistochemistry 1:100,000, Immunocytochemistry/Immunofluorescence 1:50 | |
| MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein | |
| Rat | |
| Protein G purified | |
| Hypoxia Signaling, Immune System Diseases, Immunology, Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Phospho Specific, Stem Cell Markers | |
| 4155 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG2b |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot | |
| MBP14 | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| MBP | |
| Reacts with human MBP, recognizing epitopes in the 36-50 amino acid region (GILDSIGRFFGGDRG) | |
| 0.025 mg | |
| Primary | |
| Bovine, Chicken, Equine, Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction