missing translation for 'onlineSavingsMsg'
Learn More

MBP Antibody (7D2), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications

£163.00 - £389.00

Specifications

Antigen MBP
Clone 7D2
Concentration 1 mg/ml
Dilution Western Blot 1:5000-1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000
Applications Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18224124
View Documents
Novus Biologicals
NBP1-05204-0.025ML
0.025 mL
£163.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18738553
View Documents
Novus Biologicals
NBP1-05204
0.1 mL
£389.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

MBP Monoclonal specifically detects MBP in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

MBP
1 mg/ml
Western Blot, Immunofluorescence, Immunocytochemistry, Immunohistochemistry
Unconjugated
Mouse
Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
MBP
IgG1
Affinity purified
The MBP Antibody (7D2) antibody binds only the 21.5kDa and 18.5kDa rat MBP isotypes, but all four isotypes of human and bovine MBP.
7D2
Western Blot 1:5000-1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000
Monoclonal
Ascites
RUO
Rat, Human, Mouse, Equine, Pig, Bovine
4155
Purified myelin basic protein isolated from bovine brain, epitope maps to the peptide AEGQRPGFGYGGRASDYKSAHKGFKGVDAQGTLSKIFKLG, amino acids 145-184 of the human 21.5kDa sequence.
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
18.5/21.5 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.