missing translation for 'onlineSavingsMsg'
Learn More

MBP Antibody (7D2), Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications

Brand:  Novus Biologicals NBP1-05204

 View more versions of this product

Product Code. 18738553

  • £389.00 / 0.10mL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MBP Monoclonal specifically detects MBP in Human, Mouse, Rat, Porcine, Bovine, Equine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MBP
Monoclonal
1 mg/ml
Western Blot 1:5000-1:10000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:1000
MGC99675, Myelin A1 protein, myelin basic protein, Myelin membrane encephalitogenic protein
Mouse
18.5/21.5 kDa
0.1 mL
Neuroscience, Neurotransmission, Oligodendrocyte Cell Markers, Stem Cell Markers
4155
Human, Mouse, Rat, Pig, Bovine, Equine
Ascites
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
7D2
Unconjugated
50% PBS, 50% glycerol with 5mM Sodium Azide
MBP
Purified myelin basic protein isolated from bovine brain, epitope maps to the peptide AEGQRPGFGYGGRASDYKSAHKGFKGVDAQGTLSKIFKLG, amino acids 145-184 of the human 21.5kDa sequence.
Affinity purified
RUO
Primary
The MBP Antibody (7D2) antibody binds only the 21.5kDa and 18.5kDa rat MBP isotypes, but all four isotypes of human and bovine MBP.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only