missing translation for 'onlineSavingsMsg'
Learn More

MBL™ Graduated Class B Type 1 Glass Pipets

Measure liquids with permanent amber stain graduations. MBL™ graduated class B glass pipets with the zero mark at the top (type 1) makes it easy.

Brand:  MBL PRA760B

Code : 33

Additional Details : Weight : 0.19000kg

 View more versions of this product

Product Code. 12590036

  • £16.30 / Pack of 5
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

  • Complies with ISO 835
  • Calibrated to deliver from zero at the top to any graduation line down to the shoulder
  • Graduations in permanent amber stain which withstands aggressive cleaning solutions
  • Manufactured from soda lime glass with robust tooled jets
Specifications

Specifications

To Deliver
Soda Lime Glass
0.01 mL
±0.01 mL
1 mL
ISO 835
Yes
5/Pk.
Yellow
Graduated Pipette
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers