Learn More
MAZ Rabbit, anti-Human, Clone: 1D5, Proteintech
Rabbit Recombinant Monoclonal Antibody
Brand: Proteintech 82852-3-RR-20UL
This item is not returnable.
View return policy
Description
The MAZ antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human MAZ. This antibody recognizes human antigen. The MAZ antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
| MAZ | |
| 1D5 | |
| MAZ, MAZI, Pur 1, PUR1, SAF 1, SAF 2, ZF87, Zif87, ZNF801 | |
| 20 μL | |
| 4150 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Immunofluorescence, Immunohistochemistry, ELISA | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG |