missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAZ Rabbit, anti-Human, Clone: 1D5, Proteintech
Rabbit Recombinant Monoclonal Antibody
£135.00 - £360.00
Specifications
| Antigen | MAZ |
|---|---|
| Clone | 1D5 |
| Concentration | 1000 μg/mL |
| Applications | Immunofluorescence, Immunohistochemistry, ELISA |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18084225
|
Proteintech
82852-3-RR-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18020706
|
Proteintech
82852-3-RR-100UL |
100 μL |
£360.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MAZ antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human MAZ. This antibody recognizes human antigen. The MAZ antibody has been validated for the following applications: IF, IHC, ELISA analysis.
Specifications
| MAZ | |
| 1000 μg/mL | |
| Liquid | |
| Human | |
| MAZ, MAZI, Pur 1, PUR1, SAF 1, SAF 2, ZF87, Zif87, ZNF801 | |
| Primary |
| 1D5 | |
| Immunofluorescence, Immunohistochemistry, ELISA | |
| Rabbit | |
| 4150 | |
| IgG | |
| Store at -20°C. Stable for one year after shipment. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title