Learn More
3M™ Maxim Series Polycarbonate Lens Safety Goggles
Product Code. 12632545
Change view
Click to view available options
Lens Tint:
Amber
Bronze
Clear
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Lens Tint unitSize
12632545 Clear Each
12622605 Bronze Each
12632615 Amber Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12632545 Supplier 3M™ Supplier No. 7010045682
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Specifications
Certifications/Compliance EN 166: 2001
Lens Coating Anti-fog, Scratch-resistant
Lens Tint Clear
Product Type Safety goggles
For Use With (Application) Construction
Frame Color Black
No. per Pack 1 Ea.
Material Polycarbonate