Learn More
3M™ Maxim™ RAS Safety Glasses
Product Code. 15760028 Shop All 3M Products
Change view
Click to view available options
Lens Tint: Clear
Product Code.
15760028 Lens Tint: Clear
1 options
Product Code. 15760028 Supplier 3M™ Supplier No. 7100010414
Quantity Price
1 £213.57 / Pack of 20
6 £202.85 / Pack of 20
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
missing translation for 'validMainframeMsg'
3M™ Maxim™ RAS Safety Glasses provides excellent protection against scratches in the toughest of environments.
  • Safety spectacles with a super hard eyewear coating that provides excellent protection against scratches in the toughest of environments
  • RAS coating 5 times more scratch resistance than generic hard coats
  • RAS coating fulfils the EN 166 requirements for K marking
  • Spectacles with ventilation channel across the lens to minimize misting
  • Soft, adjustable nose bridge and a soft-touch brow guard
  • Adjustable temple lengths and pantoscopic (angle) adjustments
  • Design provides excellent coverage and good field of vision
TRUSTED_SUSTAINABILITY
Specifications
Lens Coating Scratch-resistant
Lens Tint Clear
Product Type Safety glasses
Frame Color Black