Learn More
3M™ Maxim™ DX Ballistic Safety Glasses
Product Code. 15769657
Click to view available options
Lens Tint:
Amber
Bronze
Clear
Unit Size:
Pack of 20
Product Code. 15769657
Brand: 3M™ 7010045673
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
3M™ Maxim™ DX Ballistic Safety Glasses are designed specifically to wear in combination with a helmet and or ear muffs.
  • Safety spectacles that exceed the Military VO test regimen for high velocity impact
  • DX coating provides enhanced anti-scratch and anti-fog protection
  • Spectacles with ventilation channel across the lens to minimize misting
  • Soft, adjustable nose bridge and a soft-touch brow guard
  • Adjustable temple lengths and pantoscopic (angle) adjustments
  • Design provides excellent coverage and good field of vision
  • Low-profile design increases compatibility with helmets and hearing protection
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN 166:2001
Lens Type DX
Lens Tint Clear
Product Type Ballistic Safety Glasses
For Use With (Application) Construction
Frame Color Black, Gray
Lens Material Polycarbonate