missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAVS; VISA Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
£119.00 - £330.00
Specifications
| Antigen | MAVS; VISA |
|---|---|
| Clone | 7F14 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30235818
|
Proteintech
81910-1-RR-100UL |
100 μL |
£360.00 £330.00 / 100µL Save £30.00 8% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233388
|
Proteintech
81910-1-RR-20UL |
20 μL |
£135.00 £119.00 / 20µL Save £16.00 11% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The MAVS; VISA antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human MAVS; VISA. This antibody recognizes human antigen. The MAVS; VISA antibody has been validated for the following applications: WB, IHC, IF/ICC, IP, ELISA analysis.Specifications
| MAVS; VISA | |
| 1000 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 57506 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 7F14 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human | |
| CARDIF; DKFZp547C224; DKFZp666M015; FLJ27482; FLJ35386; FLJ38051; FLJ41962; IPS-1; IPS1; KIAA1271; MGC3260; VISA | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title