missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAVS; VISA Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
Brand: Proteintech 81910-1-RR-20UL
This item is not returnable.
View return policy
Description
The MAVS; VISA antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human MAVS; VISA. This antibody recognizes human antigen. The MAVS; VISA antibody has been validated for the following applications: WB, IHC, IF/ICC, IP, ELISA analysis.Specifications
| MAVS; VISA | |
| Recombinant Monoclonal | |
| 1000 μg/mL | |
| CARDIF; DKFZp547C224; DKFZp666M015; FLJ27482; FLJ35386; FLJ38051; FLJ41962; IPS-1; IPS1; KIAA1271; MGC3260; VISA | |
| Recombinant Protein | |
| 20 μL | |
| Primary | |
| Human | |
| 75 kDa | |
| Liquid |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, ELISA | |
| 7F14 | |
| Unconjugated | |
| Rabbit | |
| Protein A purification | |
| RUO | |
| 57506 | |
| Store at -20°C. Stable for one year after shipment. | |
| Recombinant Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction