missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MATK Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
£587.00
Specifications
| Antigen | MATK |
|---|---|
| Dilution | Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
MATK Polyclonal antibody specifically detects MATK in Human samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
| MATK | |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| Cell Cycle and Replication | |
| PBS, pH 7.2, 40% glycerol | |
| 4145 | |
| IgG | |
| Affinity purified |
| Western Blot 0.04-0.4 ug/ml, Immunocytochemistry/Immunofluorescence 0.25-2 ug/ml | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| CHK, Csk-type protein tyrosine kinase, CTKEC 2.7.10.2, DKFZp434N1212, EC 2.7.10, Hematopoietic consensus tyrosine-lacking kinase, HHYLTK, hydroxyaryl-protein kinase, HYL tyrosine kinase, HYLCsk-homologous kinase, HYLTK, leukocyte carboxyl-terminal src kinase related, Lsk, megakaryocyte-associated tyrosine kinase, megakaryocyte-associated tyrosine-protein kinase, MGC1708, MGC2101, Protein kinase HYL, tyrosine kinase MATK, Tyrosine-protein kinase CTK, tyrosylprotein kinase | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: YSKDKGAICTKLVRPKRKHGTKSAEEELARAGWLLNLQHLTLGAQIGEGEFGAVLQGEYLGQKVAVKNIKC | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title