missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAT1/2A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
£161.00 - £386.00
Specifications
| Antigen | MAT1/2A |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 2 ug/ml, Simple Western 1:10 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18204180
|
Novus Biologicals
NB110-94162SS |
0.025 mL |
£161.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18201500
|
Novus Biologicals
NB110-94162 |
0.1 mL |
£386.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MAT1/2A Polyclonal specifically detects MAT1/2A in Human, Rat, Bovine, Primate, Zebrafish, Mouse (Negative) samples. It is validated for Western Blot, Simple Western.Specifications
| MAT1/2A | |
| Western Blot 2 ug/ml, Simple Western 1:10 | |
| Polyclonal | |
| Rabbit | |
| Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Signal Transduction | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| AdoMet synthase 2, AMS2, EC 2.5.1.6, MAT 2, MATA2SAMS2adoMet synthetase 2, MATII, MAT-II, Methionine adenosyltransferase 2, Methionine adenosyltransferase II, methionine adenosyltransferase II, alpha, S-adenosylmethionine synthase isoform type-2, S-adenosylmethionine synthetase isoform type-2 | |
| MAT2A | |
| IgG | |
| Affinity Purified | |
| Human, rat, orangutan, bovine, Zebrafish and monkey. Does not react with mouse. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human, Rat, Bovine, Primate, Zebrafish, Mouse (Negative) | |
| P31153 | |
| 4144 | |
| Synthetic peptide made to an internal portion of the human MAT2A protein (within residues 100-200). [Swiss-Prot# P31153] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 43 kDa |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title