missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAT1/2A Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NB110-94162
This item is not returnable.
View return policy
Description
MAT1/2A Polyclonal specifically detects MAT1/2A in Human, Rat, Bovine, Primate, Zebrafish, Mouse (Negative) samples. It is validated for Western Blot, Simple Western.
Specifications
| MAT1/2A | |
| Polyclonal | |
| Unconjugated | |
| PBS and 30% Glycerol with 0.1% Sodium Azide | |
| AdoMet synthase 2, AMS2, EC 2.5.1.6, MAT 2, MATA2SAMS2adoMet synthetase 2, MATII, MAT-II, Methionine adenosyltransferase 2, Methionine adenosyltransferase II, methionine adenosyltransferase II, alpha, S-adenosylmethionine synthase isoform type-2, S-adenosylmethionine synthetase isoform type-2 | |
| Rabbit | |
| 43 kDa | |
| 0.1 mL | |
| Amino Acids Drugs and other small molecules, Cancer, Lipid and Metabolism, Signal Transduction | |
| 4144 | |
| Human, Rat, Bovine, Primate, Zebrafish, Mouse (Negative) | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 2 ug/ml, Simple Western 1:10 | |
| P31153 | |
| MAT2A | |
| Synthetic peptide made to an internal portion of the human MAT2A protein (within residues 100-200). [Swiss-Prot# P31153] | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, rat, orangutan, bovine, Zebrafish and monkey. Does not react with mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction