missing translation for 'onlineSavingsMsg'
Learn More

Masterflex™ Stainless Steel Thermoset Polyester Housing High-Performance Pump Head

Product Code. 11711209
Click to view available options
Flow Rate:
0.01 to 19 L/min.
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 11711209

Brand: Masterflex™ 7760062

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mount this pump head with tubing exiting to the top or side for flexible setup. Masterflex™ I/P™ High-Performance Pump Head offers the highest flow rates, best pressure and suction performance of any I/P™ pump head.

  • C-shaped design allows tubing to enter and exit the same side of the pump head.
  • Polyester and stainless steel construction withstands tough process environments
  • Versatile mounting configurations
  • Facilitates connection to tubing inlet and outlet
  • Adjustable tubing retention prevents tubing movement in head

Specifications

Flow Rate 0.01 to 19 L/min.
Model 77600-62
Occlusion Type Fixed
Roller Material Stainless Steel
Tubing Type I/P 70, I/P 88, I/P 89
Rotor Material Stainless Steel
Speed 1 to 650 rpm
Case Material Stainless Steel, Thermoset Polyester
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.