Learn More
Masterflex™ Mounting Kit
Product Code. 11822603
Click to view available options
Material:
Stainless Steel
Unit Size:
Each
Product Code. 11822603
Brand: Masterflex™ 7780000
Only null left
Add to basket
Add to basket
Easy stack multiple pump heads on your Masterflex™ L/S™ drive
  • Constructed of durable stainless steel
  • See drive specifications for recommended number of heads
  • Suitable for use with one Easy-Load™ 3 pump head
Specifications
For Use With (Application) L/S Easy-load 3 Pump Head Connection
For Use With Masterflex L/S Pump Drives
Includes Single-channel mounting hardware and a 15 in. length of Silicone pump tubing
Material Stainless Steel
No. of Channels 1
Quantity 1 Each
Type Mounting Kit