Learn More
Masterflex™ I/P™ End Bell Assembly
Complete your pump head assembly with two end bell assemblies. MasterFlex™ I/P™ End Bell Assemblies are designed for use with Masterflex™ I/P™ 73 Standard Pump Heads.
Brand:  Masterflex™ 07019-79
Code : A0
Additional Details : Weight : 0.25200kg
 View more versions of this product
Product Code. 11786068
  • £322.00 / Each
Estimated Shipment: 06-06-2024
to see stock.
Add to basket
Description
Description
  • Assemblies are made from high-performance Polyphenylene sulfide and stainless steel
Specifications
Specifications
For Masterflex I/P 73 standard pump heads
Polycarbonate, Stainless Steel
1 Each
End Bell Assembly