missing translation for 'onlineSavingsMsg'
Learn More

Eppendorf™ Mastercycler™ nexus X2e, 230 V/50 – 60 Hz (GB)

Product Code. 15328276 Shop All Eppendorf Products
Change view
Click to view available options
Block Configurations:
0.2/0.5 mL Tubes
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Block Configurations Unit Size
15328276 0.2/0.5 mL Tubes Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15328276 Supplier Eppendorf™ Supplier No. 6339000040
Equipment and Instrument End of Year Sale
Use promo code 28000
View all promotions
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

If you’ve got a high PCR workload, turn to the Eppendorf Mastercycler nexus X2. With two blocks (32-well and 64-well), you can run two independent protocols at the same time using a variety of consumables up to 0.5 mL tubes. For higher throughput, network with up to 3 additional family members.

Mastercycler™ nexus X2e, 230 V/50 – 60 Hz (GB)- Asymmetrically divided block for running different assays in parallel- Universal block for strips, 0.1 mL, 0.2 mL and 0.5 mL PCR tubes- Small footprint- Without gradient- E-mail notification- flexlid™ concept allows use of all types of consumables with automatic height adjustment of the lid
TRUSTED_SUSTAINABILITY

Specifications

Thermal Accuracy ±0.2°C
Thermal Uniformity ≤±0.3°C
Dimensions 41.2 x 25 x 33 cm
Capacity 64/32 x 0.2 mL PCR Tubes or up to 45/19 x 0.5 mL PCR Tubes
Voltage 230 V
Electrical Requirements 230 V, 50/60 Hz
Product Type PCR
Thermal Range 4°C to 99°C
For Use With (Application) Standard PCR, Optimizing a PCR
Frequency 50/60 Hz
User Interface Intuitive
Wattage 700 W
Max. Ramp Rate 3°C/s (Measured at the Block)
No. of Programs >700
Heated Lid Yes
Weight 11 kg
Block Configurations 0.2/0.5 mL Tubes
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.